Learn More
Poulten Selfe™ Glass Reverse Flow Viscometers
Suitable for measuring opaque liquids
256.00 €
Specifications
Product Type Reverse Flow Viscometer
View More Specs
Products 11
Product Code Brand Viscosity Range Price Quantity & Availability  
Product Code Brand Viscosity Range Price Quantity & Availability  
13141083
View Documents
Poulten Selfe™
1641/01
0.4 to 2 cSt
256.00 €
Each
13151083
View Documents
Poulten Selfe™
1641/02
0.8 to 4 cSt
256.00 €
Each
13141093
View Documents
Poulten Selfe™
1641/11
1600 to 8000 cSt
256.00 €
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
13161083
View Documents
Poulten Selfe™
1641/03
1.6 to 8 cSt
256.00 €
Each
13111093
View Documents
Poulten Selfe™
1641/08
100 to 500 cSt
256.00 €
Each
13191083
View Documents
Poulten Selfe™
1641/06
20 to 100 cSt
256.00 €
Each
13121093
View Documents
Poulten Selfe™
1641/09
240 to 1200 cSt
256.00 €
Each
13171083
View Documents
Poulten Selfe™
1641/04
3 to 15 cSt
256.00 €
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
13151093
View Documents
Poulten Selfe™
1641/12
4000 to 20,000 cSt
256.00 €
Each
13131093
View Documents
Poulten Selfe™
1641/10
500 to 2500 cSt
256.00 €
Each
13181083
View Documents
Poulten Selfe™
1641/05
7 to 35 cSt
256.00 €
Each
Estimated Shipment: 01-05-2025
to see stock.
Add to basket
Description
Description
Opaque 240 to 1,200v(mm2s-1,cst) 400 Poulten Selfe and Lee
Specifications
Specifications
Reverse Flow Viscometer
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Poulten Selfe™ Glass Reverse Flow Viscometers