Learn More
Cole-Parmer™ Glass Sample Vials
Glass Sample Vials maintain PH level and eliminate ion leaching.
207.00 € - 488.00 €
Description
- Vials are manufactured from 33 expansion borosilicate clear glass (Type 1, Class A).
- Choose from a polypropylene cap with a PTFE-lined foam backing or a polypropylene cap with PTFE-lined silicone septa.
- Vials, Caps, and septa are shrink-wrapped separately.
Specifications
Borosilicate Glass (33 Expansion) | |
8 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Cole-Parmer™ Glass Sample Vials