missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten Selfe™ Glass Ubbelohde
Suitable for transparent liquids
227.00 € - 232.00 €
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
13161063
|
Poulten Selfe™
1643/06 |
1B |
232.00 €
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
13191063
|
Poulten Selfe™
1643/09 |
2B |
232.00 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
||||||
13101073
|
Poulten Selfe™
1643/10 |
3 |
232.00 €
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
13111073
|
Poulten Selfe™
1643/11 |
3C |
232.00 €
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
13131073
|
Poulten Selfe™
1634/13 |
4 |
227.00 €
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
Description
ASTM D445-IP71, ASTM D446. Suitable for transparent liquids. Nominal overall length 283mm. Approximate sample size 18mL. With UKAS certificate of calibration.Specifications
Ubbelohde Viscometer | |
Yes | |
Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Poulten Selfe™ Glass Ubbelohde