missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Filtrate Flask

Vacuum-resistant filtrate flask for the Sartorius™ All-Glass Vacuum Filter Holder system. Sartorius™ Filtrate Flask is constructed of durable borosilicate glass 3.3.

Brand:  Sartorius™ 6983001

202.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10161541

  • 289.00 € / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Vacuum resistant
  • Can be used to retain filtrate
  • A 6 mm wide, non-ground glass rim, located above the ground glass neck of the filtrate flask prevents contamination of the filtrate by contact with the ground glass area (e.g., by grease) while it is being poured out.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1000 mL
Borosilicate Glass (3.3)
All-glass vacuum filter holder assembly
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius™ Filtrate Flask > Product Type: Filtrate Flask

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.