missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GlutaMAX™ Supplement

GlutaMAX™ Supplement
Brand: Gibco 35050038
33.19 EUR valid until 2025-12-31
Use promo code "24071" to get your promotional price.
This item is not returnable.
View return policy
Description
Gibco™ GlutaMAX™ Supplement is an alternative to L-glutamine, with increased stability that improves cell health. GlutaMAX™ Supplement is suitable for both adherent and suspension culture of mammalian cells, with no adaptation required. GlutaMAX™ Supplement is offered as a 200 mM L-alanyl-L-glutamine dipeptide in 0.85% NaCl. GlutaMAX™ Supplement is also included in a variety of media formulations. Compared to L-glutamine, GlutaMAX™ Supplement:Features:
Minimizes toxic ammonia build-up
Unlike L-glutamine, GlutaMAX™ Supplement does not spontaneously break down to form ammonia. Instead, cells cleave the dipeptide bond to release L-glutamine as needed. This system prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.
Improves cell viability and growth
Cultures with lower levels of ammonia show improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of GlutaMAX™ Supplement that is not possible with L-glutamine.
Remains stable across a wide range of temperatures
GlutaMAX™ Supplement is room-temperature stable and ready to use. GlutaMAX™ Supplement retains full functionality for 24 months at room temperature, 2 to 8°C, or -5 to -30°C for convenient storage and handling. GlutaMAX™ Supplement is even stable at 37°C for a week or more.
cGMP manufacturing and quality system
Gibco™ GlutaMAX™ supplement is manufactured at a cGMP compliant facility, located in Paisley, Scotland. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ GlutaMAX™ supplement made in our Grand Island, New York facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Specifications
cGMP, ISO 13485 standard, FDA | |
GlutaMAX™ Supplement, 100mL, 100X Concentration, Liquid Form, Suitable for Both Adherent and Suspension Culture of Mammalian Cells, with No Adaptation Required, GlutaMAX™ Supplement is Offered as a 200mM L-Alanyl-L-Glutamine Dipeptide in 0.85% | |
Liquid | |
Mammalian Cell Culture | |
For in-vitro diagnostic use |
15 to 30°C | |
100 X | |
GlutaMax | |
100 mL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GlutaMAX™ Supplement > 100mL
Spot an opportunity for improvement?Share a Content Correction