missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ N-2 Supplement (100X)

N-2 Supplement (100X)

Brand:  Gibco™ 17502048

91.91 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 11520536

  • 101.00 € / 5mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

N-2 Supplement is a chemically defined, serum-free supplement based on Bottenstein’s N-1 formulation.

N-2 Supplement is recommended for growth and expression of neuroblastomas as well as post-mitotic neurons in primary cultures from both the peripheral nervous system (PNS) and the central nervous system (CNS). N-2 Supplement can be used as a substitute for Bottenstein’s N-1 formulation. N-2 Supplement is provided as a 100X concentrate.

N-2 Supplement may be used with Neurobasal™ media supplemented with growth factors such as bFGF and EGF or used with DMEM.

For Research Use Only. Not intended for any animal or human therapeutic of diagnostic use.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -20°C) and protect from light.

Serum-free
Mammalian Cell Culture
Cell Culture Supplement
Sterile
5 mL
Endotoxin, Performance, Sterility
Neural Cells
100 X
Liquid
Serum-Free
6 to 8
Dry Ice
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Mercuric acetate

Supplemental information
  • MIXTURE LIST-Contains : Mercuric acetate

SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ N-2 Supplement (100X) > 5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.