missing translation for 'onlineSavingsMsg'
Learn More
Glycine, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10101620

Glycine, Certified AR for Analysis, Fisher Chemical™

Product Code. 10101620
100 g
Click to view available options
Quantity:
100 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 10101620

Brand: Fisher Chemical G/0800/48

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C2H5NO2, CAS Number-56-40-6, glycolixir, padil, aciport, hampshire glycine, aminoethanoic acid, glycine, glycocoll, glycosthene, glicoamin, aminoacetic acid, 1kg, CHEBI:15428, White, 75.07g/mol, DHMQDGOQFOQNFH-UHFFFAOYSA-N, 2-aminoacetic acid, 182 deg.C, 8131, 75.067, Plastic powder jar, 5.9 to 6.4

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point 182°C
pH 5.9 to 6.4
Color White
Packaging Plastic Powder Jar
Quantity 100 g
Formula Weight 75.07g/mol
Physical Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycine, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.