missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, Certified AR for Analysis, Fisher Chemical™
C2H5NO2, CAS Number-56-40-6, glycolixir, padil, aciport, hampshire glycine, aminoethanoic acid, glycine, glycocoll, glycosthene, glicoamin, aminoacetic acid, 1kg, CHEBI:15428, White, 75.07g/mol, DHMQDGOQFOQNFH-UHFFFAOYSA-N, 2-aminoacetic acid, 182 deg.C, 8131, 75.067, Plastic powder jar, 5.9 to 6.4
39.25 € - 197.00 €
Chemical Identifiers
CAS | 56-40-6 |
---|---|
Molecular Formula | C2H5NO2 |
Molecular Weight (g/mol) | 75.07 |
MDL Number | MFCD00008131 |
InChI Key | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
Synonym | glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine |
PubChem CID | 750 |
ChEBI | CHEBI:15428 |
IUPAC Name | 2-aminoacetic acid |
SMILES | NCC(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10101620
|
Fisher Chemical
G/0800/48 |
100 g |
39.25 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10589760
|
Fisher Chemical
G/0800/50 |
250 g |
75.25 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10070150
|
Fisher Chemical
G/0800/60 |
1 kg |
197.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C2H5NO2, CAS Number-56-40-6, glycolixir, padil, aciport, hampshire glycine, aminoethanoic acid, glycine, glycocoll, glycosthene, glicoamin, aminoacetic acid, 1kg, CHEBI:15428, White, 75.07g/mol, DHMQDGOQFOQNFH-UHFFFAOYSA-N, 2-aminoacetic acid, 182 deg.C, 8131, 75.067, Plastic powder jar, 5.9 to 6.4
Chemical Identifiers
56-40-6 | |
75.07 | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
750 | |
2-aminoacetic acid |
C2H5NO2 | |
MFCD00008131 | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
CHEBI:15428 | |
NCC(O)=O |
Specifications
182°C | |
5.9 to 6.4 | |
C2H5NO2 | |
Plastic Powder Jar | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
2-aminoacetic acid | |
750 | |
75.07g/mol |
56-40-6 | |
White | |
MFCD00008131 | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
NCC(O)=O | |
75.07 | |
CHEBI:15428 | |
Powder |