missing translation for 'onlineSavingsMsg'
Learn More

H-B Instrument™ Enviro-Safe™ Dry Block/Incubator Thermometers

EnviroKleen™-certified environmentally friendly with 100% non-toxic, non-hazardous, biodegradable liquid fill. H-B Instrument™ Enviro-Safe™ Dry Block/Incubator Thermometers are 175mm (6.9 in.) liquid-in-glass, 35mm immersion thermometers.

Brand:  H-B Instrument™ B60510-0100

86.32 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 13596980

  • 93.08 € / Each
Estimated Shipment: 30-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • USA made thermometers include Enviro-Safe™ green liquid against white back glass
  • EnviroKleen™-certified environmentally friendly and packaged in recyclable Safety-Pak™ packaging
  • Feature the same accuracy as similar mercury thermometers, with arrow indicators imprinted on the glass at 25, 30, 37 and 56°C
  • Individually serialized and tested in a triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory
  • Internationally recognized and accepted by the signatory economies of the ILAC/MRA
  • Multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST
  • Mfr. No. B60510-0100 features PFA flouropolymer safety coating to keep liquid and glass encapsulated in the event of breakage 
Specifications

Specifications

ISO/IEC 17025:2005, A2LA, NIST
175 mm
Enviro-Safe™ Liquid
57°C
24°C to 57°C
0.5°C
6.9 in.
Glass
Dry Block/Incubator Thermometer, PFA Coated
24°C
0.5°C
35 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
H-B Instrument™ Enviro-Safe™ Dry Block/Incubator Thermometers > PFA coated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.