missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ HACCP Flat Tray with Lid and Grid

HACCP Flat Tray can be used as a defrosting tray due to the wide dividing grid. Complete package with tray, shelf, and lid.

Brand:  Gilac™ G190128

 View more versions of this product

Product Code. 16480821

  • 18.40 € / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Flat Tray, Complete package: Tray + Shelf + Lid
  • Dimensions suitable for refrigerators
  • Can be used as:
    • defrosting trays thanks to its wide dividing grid that separates the ice water from the product being defrosted
    • preparation container for products that drip ingredients tray
    • containers for preserving comparison samples
    • fermenting and decanting containers
  • Nesting and stacking with lid
Specifications

Specifications

Food Bin
Blue
101.4 oz.
HDPE
Rectangular
3 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gilac™ HACCP Flat Tray with Lid and Grid > HACCP Flat Tray with Lid and Grid; Capacity: 3 L; Color: Blue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.