missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Aquarius™ C-Fold White Hand Towel Dispenser

Aquarius™ C-Fold White Hand Towel Dispenser promotes hygiene and helps to manage costs through controlled dispensing.

Brand:  Kimberly-Clark™ 6954

37.05 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15242896

  • 52.00 € / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Dispenses folded hand towels without the need to touch the dispenser, helping to reduce contamination and the spread of germs
  • Hassle-free maintenance keeps costs down in areas of heavy use
  • Easy to unlock and refill, with a window to gauge refill needs
  • Sleek and contemporary design with a white, high-gloss, easy-clean finish and no dirt or dust traps
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C-fold Hand Towel Dispenser
420 x 320 x 151 mm
Plastic
1
Hand towel dispenser suitable for use in washrooms or laboratory
White
Paper Towel
Wall Mount
Hinged Lid with Front Opening
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ Aquarius™ C-Fold White Hand Towel Dispenser > Color: White

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.