missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Hardened Steel Grinding Jars
Brand: RETSCH 01.462.0300
This item is not returnable.
View return policy
Specifications
Grinding Jar | |
5 mL | |
For CryoMill | |
Hardened Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RETSCH Hardened Steel Grinding Jars > Capacity: 5mL
Spot an opportunity for improvement?Share a Content Correction