Learn More
Fisherbrand™ Heat Sealer with Cutter
Seal bags, sheets or tubing quickly and efficiently. Heat Sealer with Cutter features a safe heating element that is only hot when in use. Timing knob allows for adjustable sealing duration. 450 Watt unit (220/230 VAC) includes a replacement part kit. Available with a UK or European plug.
Brand: Fisherbrand™ HS120289
Beschreibung
Seal bags, sheets or tubing quickly and efficiently. Fisherbrand™ Heat Sealer with Cutter features a safe heating element that is only hot when in use. Timing knob allows for adjustable sealing duration. 450 Watt unit (220/230 VAC) includes a replacement part kit. Available with a UK or European plug.
- Seal polyethylene or polypropylene bags, sheets or flat tubing from 0.01 to 0.8 mm thick.
- Sliding blade removes excess material 19 mm from the seal.
- Heating element measures 310 mm long and is only hot when sealing.
- Timing knob ensures safe sealing.
- Simply turn knob to set time (1-8 seconds).
- Press handle down.
- Remove when light indicates cycle is complete.
- Includes replacement part kit (additional replacement part kits can be ordered separately):
- Two extra blades
- Two extra heating elements
- Two extra PTFE cloths
Spezifikation
Commercial Pouch Sealer | |
Polyethylene or polypropylene bags, sheets or flat tubing | |
Adjustable, 1 to 8 seconds |
446 x 86 x 166 mm (closed) | |
Replacement part kit (two each of: blades, heating elements, PTFE cloths), 153 cm power cord | |
Cont. Europe/Israel Plug |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.