Learn More
Labconco™ Heated Product Shelves, Small
Heated Product Shelves consist of three aluminum shelves with 1/4 in. rims, three stainless steel support rods, nine clips for shelf height adjustment, low voltage transformer with variable heat control to +40° C (+104° F), and three rubber feet.
Brand:  Labconco™ 7509201
Product Code. 10286433
  • 4680.00 € / Pack of 3
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Heated Product Shelves consist of three aluminum shelves with 1/4 in. rims, three stainless steel support rods, nine clips for shelf height adjustment, low voltage transformer with variable heat control to +40° C (+104° F), and three rubber feet. Product Shelves accommodate bulk samples, microtiter plates, vials or serum bottles. They are designed to fit inside Drying Chambers (sold separately).
Specifications
Specifications
Heated Shelf
CE
0 to 300 V
Aluminum
Drying Chambers