Learn More
Grant Instruments™ Heating Block for TS-DW Deep-well Microplate Thermoshaker
Accommodates one deep-well plate for TS-DW Deep-well Microplate Thermoshaker
861.00 €
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
15450474
|
Grant Instruments™
B-2A |
Axygen™ 96/2200µl deep-well plate |
861.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
15430474
|
Grant Instruments™
B-2E |
Eppendorf™ 96/1000µl deep-well plate |
861.00 €
Each |
Estimated Shipment: 06-03-2025
Log in to see stock. |
|||||
15440474
|
Grant Instruments™
B-2S |
Sarstedt™ 96/2200µl deep-well plate |
861.00 €
Each |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
15460474
|
Grant Instruments™
B-2SL |
Starlab™ 96/1200µl deep-well plate |
861.00 €
Each |
Estimated Shipment: 06-03-2025
Log in to see stock. |
|||||
Description
Block for one deep-well plate- Shaped to fit the microplates for better heat transfer
- Three year warranty
Specifications
TS-DW Microplate Thermoshaker | |
Block |