missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Hei-TORQUE Expert 200

With Hei-TORQUE Value 200 you can optimally stir viscosities up to 100,000 mPas or volumes up to 50 liters.

Brand:  Heidolph™ 501-62010-03

2194.50 EUR valid until 2025-12-31
Use promo code "24303" to get your promotional price.


Product Code. 17332990

  • 2310.00 € / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With Hei-TORQUE Value 200 you can optimally stir viscosities up to 100,000 mPas or volumes up to 50 liters (H2O). Thanks to a wide speed range from 10 to 2000 rpm and a maximum torque of 200 Ncm, it works particularly powerful.
  • The laboratory stirrer Hei-TORQUE Value 200 performs every stirring task with minimal noise thanks to the latest engine technology. This laboratory stirrer is also very user-friendly in terms of operation and safety. The 2.4 inch display makes it very easy to set the application parameters and control the process (e.g. using the torque trend display). The safety-related start/stop touch-function serves to minimize the risk of unintentional switching on.
  • Suitable for use in fume hoods or as a drive for reactor super structures, the Hei-TORQUE Value 200 has a quick-action chuck (quick chuck with safety ring) with a diameter of 10.5 mm, which can be combined with a large number of stirring tools for the best stirring results.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

230 V
UK
200 Ncm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heidolph™ Hei-TORQUE Expert 200 > UK Plug

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.