Learn More
Cytiva HiTrap™ Capto™ Q ImpRes IEX Columns
High-throughput medium for intermediate and polishing protein purification
Brand:  Cytiva 17-5470-55
 View more versions of this product
Product Code. 11673074
  • 384.00 € / Pack of 5
Explore more special offers
Description
Description
  • Combined properties of high binding capacity, high flow rate and low back pressure reduces process cycle times and improves productivity at process scale
  • Rigid agarose matrix enables high bed heights and purification of viscous samples at high flow rates
  • Screening of selectivity, binding and elution conditions, as well as small scale purifications
  • Strong quartenary amine anion exchanger
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HiTrap Capto Q ImpRes, 5 x 5 mL
HiTrap Capto Q ImpRes chromatography column is packed with a strong anion exchange modern resin for high resolution, small-scale protein purification.
7 mm
5 x 5 mL
<20 mL/min.
4°C to 30°C in 20% Ethanol
5 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva HiTrap™ Capto™ Q ImpRes IEX Columns > HiTrap Capto Q ImpRes prepacked column; Pack size: 5 x 5mL