missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Hollow Cathode Lamp, Calcium, 37mm

Achieve reliable atomic absorption results with high-performance lamps for your spectrometers

Brand:  Cole-Parmer™ 11949-63

Product Code. 13033540

  • 385.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Calcium (Ca) single-element hollow cathode lamps are quick to reach equilibrium performance, offer a long-term constant light intensity, and low noise with good calibration. Inside each lamp, there is a high volume of inert gas to increase the lamp's lifetime to 5000 mA hours.

  • Carry out quicker start-up times
  • Reduce your operating costs with longer lifetime amp hours
  • Continuous light intensity for longer periods of time
  • Compatible with: Uncoded
Specifications

Specifications

Calcium
11949-63
Hollow Cathode Lamp
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Hollow Cathode Lamp, Calcium, 37mm >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.