Learn More
Invitrogen™ Human MDR1 Vesicles
Human MDR1 Vesicles are insect derived purified plasma membranes with inserted MDR1 transport protein (P-glycoprotein, ABCB1). Our Human MDR1 Vesicles can be used to evaluate test compounds and drug interactions with MDR1 in in vitro assays.
Brand: Invitrogen™ GM0015
Description

Specifications
Frozen | |
Research Grade | |
Human Vesicle | |
Vial |
0.5 mL | |
Human | |
Store at -80°C. | |
Dry Ice |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.