Learn More
Invitrogen™ Human RGPD5 Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP0686
Product Code. 15348890
  • 141.00 € / 50µg
Explore more special offers
Description
Description
RGPD5 Synthetic Peptide, PEP-0686, from Invitrogen.
The RANBP2-like and GRIP domain containing 5 protein (RGPD5) has high similarity to RANBP2, a large RAN-binding protein localized at the cytoplasmic side of the nuclear pore complex. The gene coding for RGPD5 is thought to have arisen from a gene duplication event of RANBP2 as these highly homologous genes are located close to each other at chromosome 2q11-q12. RGPD5 was identified as an HIV dependency factor (HDF), suggesting that RGPD5 may be an important drug target in HIV treatment. At least two isoforms of RGPD5 are known to exist, of which the shorter isoform is expressed primarily in testis, while the longer of the two is expressed at low levels in a number of somatic tissues.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
84220
purified
Human RGPD5
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q99666
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
RGPD5
0.02% sodium azide
50 μg
PBS
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Human RGPD5 Synthetic Peptide > 50 μg; Unlabeled