Learn More
Cytiva HyClone™ CDM4MAb Powdered Cell Culture Media
ADCF medium for use with hybridoma and myeloma cell lines.
Brand: Cytiva SH30800.01
Description
HyClone CDM4MAb is a chemically defined and animal-derived component-free (ADCF) medium. This regulatory-friendly medium is designed to increase process yields for the manufacture of antibodies and antibody fragments for therapeutic use in a variety of engineered hybridoma and myeloma cell lines.
- Animal-derived component-free.
- Chemically defined formulation
- Designed for high cell yield and monoclonal antibody (MAb) production.
- Allows for direct or sequential adaptation
- Manufactured from traceable components according to cGMP guidelines
Specifications
2 to 8°C, away from light and moisture | |
Non-sterile | |
5 L |
Powder | |
No Glutamine, No Poloxamer 188 |
Product Suggestions
Customers who viewed this item also viewed