missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cryopreservation Media
Animal origin-free cryopreservation media for general use
Brand: Cytiva SR30001.02
This item is not returnable.
View return policy
Description
- Serum-free, animal origin-free for cryogenic storage for standard cell lines
- Meets industry standard for viable cell recovery per vial; outperforms other cryo media solution in the market for viable cell recovery per vial
- Provided as a 2X solution; 0.2μm sterile filtered
- For further manufacturing or research use. Not for diagnostic or therapeutic use.

Specifications
100 mL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Cryopreservation Media > HyCryo Media; 100mL
Spot an opportunity for improvement?Share a Content Correction