missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Solution
Buffer cell culture with PBS in solution.
18.40 € - 520.00 €
Specifications
Certifications/Compliance | Manufactured under cGMP and ISO 9001 certified facilities |
---|---|
Content And Storage | 15° to 30°C |
Form | Liquid |
Product Type | Dulbecco's Phosphate Buffered Saline |
Sterility | Sterile |
Product Code | Brand | Quantity | Concentration | With Additives | Without Additives | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Concentration | With Additives | Without Additives | Price | Quantity & Availability | ||
11581811
|
Cytiva
SH30028.01 |
100 mL | 1 X | None | No Calcium, No Magnesium, No Phenol Red |
18.40 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10687974
|
Cytiva
SH30028.LS |
6 x 1000 mL | 1 X | None | No Calcium, No Magnesium, No Phenol Red |
212.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10540853
|
Cytiva
SH30264.02 |
1000 mL | 1 X | Calcium, Magnesium | No Phenol Red |
32.40 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11591811
|
Cytiva
SH30028.08 |
10 L | 1 X | None | No Calcium, No Magnesium |
471.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11511821
|
Cytiva
SH30028.FS |
6 x 500 mL | 1 X | None | No Calcium, No Magnesium, No Phenol Red |
148.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10044823
|
Cytiva
SH30264.FS |
6 x 500 mL | 1 X | Calcium, Magnesium | No Phenol Red |
141.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10708144
|
Cytiva
SH30028.03 |
1000 mL | 1 X | None | No Calcium, No Magnesium, No Phenol Red |
35.78 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10798134
|
Cytiva
SH30264.01 |
500 mL | 1 X | Calcium, Magnesium | No Phenol Red |
24.40 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10317052
|
Cytiva
SH30028.02 |
500 mL | 1 X | None | No Calcium, No Magnesium, No Phenol Red |
24.13 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12882946
|
Cytiva
SH30378.03 |
1 L | 10 X | None | No Calcium, No Magnesium, No Phenol Red |
34.00 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10021352
|
Cytiva
SH30378.02 |
500 mL | 10 X | None | No Calcium, No Magnesium, No Phenol Red |
38.20 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11586511
|
Cytiva
SH30028.04 |
20 L | 1 X | None | No Calcium, No Magnesium |
520.00 €
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Format: Liquid
Specifications
Manufactured under cGMP and ISO 9001 certified facilities | |
Liquid | |
Sterile | |
≤0.025EU/mL | |
276±10mOsm/kg H2O | |
RUO |
15° to 30°C | |
Dulbecco's Phosphate Buffered Saline | |
0.1 μm Sterile-filtered | |
Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time | |
7 to 7.6 |
Safety and Handling
missing translation for 'shelfLife' : 24 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Solution