missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Porcine Serum (New Zealand)
Used to enhance cell culture performance, cost-efficient alternatives to Fetal Bovine Serum (FBS)
160.00 € - 167.28 €
Specifications
Country of Origin | New Zealand |
---|---|
Product Type | Porcine Serum |
Sterilization Method | Sequential Triple-filtered, 200 nm |
Endotoxin Concentration | ≤ 100 EU/mL |
Heat Inactivated | No |
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||||
16695785
|
Cytiva
SH30908.04NI |
1 L | Non-irradiated | N/A | N/A | N/A | |||||
16685785
|
Cytiva
SH30908.04 |
1 L | Irradiated | N/A | N/A | N/A | |||||
17236916
|
Cytiva
SH30908.04SAM |
1 L (QC Samples) | Irradiated | N/A | N/A | N/A | |||||
Description
Cytiva's porcine serum is sourced from New Zealand, filtered in the U.S. through multiple 200 nm pore size-rated filters and is gamma irradiated at 25 to 40 kGy after filtration.Specifications
New Zealand | |
Sequential Triple-filtered, 200 nm | |
No | |
HyClone™ |
Porcine Serum | |
≤ 100 EU/mL | |
≤ 50 mg/dL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Porcine Serum (New Zealand)