missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - NEXTGEN Coulomat C-FA, Honeywell Fluka™

Catholyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, Acetonitrile-based, free of alcohols (free of CMR substances).

Brand:  Honeywell Fluka 34470-50ML


 View more versions of this product

Product Code. 17584946

  • 415.68 € / 50mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • HYDRANAL™ is the most trusted brand in Karl Fischer titration because of its dedication to quality, consistency and continued innovation.
    • Comprehensive product portfolio with 60 available products
    • 40 years of innovation – check our NEXTGEN reagents HERE
    • Dedicated technical service and support
    • More than 800 Application Notes available on our website
    TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™ - NEXTGEN Coulomat C-FA
0.906 g/cm3
50 mL
5.5
<10.00, <70.00, ≤20.00
Liquid
Soluble in water
Orange
12°C to 13°C
75-05-8, Confidential Stabilizer 2, 75-75-2, Confidential Stabilizer 1
≥10.00, ≥60.00, ≥2.00, ≥2.00
Ampoule
UN2924
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HYDRANAL™ - NEXTGEN Coulomat C-FA, Honeywell Fluka™ > NEXTGEN Coulomat C-FA; Quantity: 50 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.