Learn More
HYDRANAL™ - NEXTGEN Solvent FI, Honeywell Fluka™
Medium for volumetric, two-component Karl Fischer titrations
Brand: Honeywell Fluka 34432-1L
View more versions of this product
Description
- Methanol-based
- Free of Imidazole
- Free of CMR Substances

Specifications
HYDRANAL™ - NEXTGEN Solvent FI | |
11°C | |
147-85-3 | |
≥0.10% | |
Glass Bottle | |
UN1230 |
0.9 g/cm3 (at 20°C) | |
1 L | |
5 to 6 | |
≤5% | |
Liquid | |
Completely Miscible in Water |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- HYDRANAL NEXTGEN Solvent FI
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol, 2-ethylimidazole, sulphur dioxide, l-proline
Your input is important to us. Please complete this form to provide feedback related to the content on this product.