Learn More
Hydrochloric acid, 0.1 N standard solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 124200010
29.80 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solution
Specifications
Hydrochloric acid | |
0.1 N Standard Solution | |
100.0°C | |
1.0000g/mL | |
1 | |
ClH | |
Liquid | |
04,450; 05,533; 06,283 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.46 |
Colorless | |
7647-01-0, 7732-18-5 | |
0.0998 to 0.1002N (20°C) | |
Glass bottle | |
1 L | |
HCl | |
MFCD00011324 MFCD00792839 | |
15, 4818 | |
Solubility in water: miscible. Other solubilities: soluble in alcohol and benzene | |
Cl | |
36.46 | |
CHEBI:17883 |
Safety and Handling
- Hydrochloric acid, 0.1 N standard solution
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : Hydrochloric acid
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0402548 | Certificate of Analysis | 21/02/2025 | 124200025,124200010,10083943,124200100,124200250 |
A0468248 | Certificate of Analysis | 12/02/2025 | 124200025,124200010,10083943,124200100,124200250 |
A0468249 | Certificate of Analysis | 12/02/2025 | 124200025,124200010,10083943,124200100,124200250 |
B00Z1692 | Certificate of Analysis | 11/06/2024 | 124200025,124200010,124200100,124200250 |
A0463221 | Certificate of Analysis | 27/05/2024 | 124200025,124200010,124200100,124200250 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only