missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, meets analytical spec. of BP, NF, Ph. Eur., fuming, 36.5-38%, Honeywell Fluka™
Shop All Honeywell Fluka Products

Click to view available options
Quantity:
1 L
2.5 L
30 kg
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Paper / Poly Bag
Plastic Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Color | Yellow |
Concentration | 36.5 to 38% |
Density | 1.2g/mL at 25°C |
Packaging | Plastic Bottle |
Residue after Ignition | ≤0.002% (as SO4) |
Vapor Pressure | 3.23psi (21.1°C), 7.93psi (37.7°C) |
Quantity | 2.5 L |
UN Number | 1789 |
Beilstein | 1098214 |
IUPAC Name | hydrogen chloride |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N2200 | Certificate of Analysis | 04/06/2024 | 07102-2.5L |
M2170 | Certificate of Analysis | 20/02/2024 | 07102-2.5L |
M2800 | Certificate of Analysis | 08/12/2023 | 07102-2.5L |
N0860 | Certificate of Analysis | 08/06/2023 | 07102-2.5L |
M3400 | Certificate of Analysis | 14/04/2023 | 07102-2.5L |
Safety and Handling


Product Identifier
- Hydrochloric acid
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrochloric acid, meets analytical spec. of BP, NF, Ph. Eur., fuming, 36.5-38%, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction