missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, for analysis, fuming, 37% solution in water, Thermo Scientific Chemicals

23.10 € - 151.00 €

Chemical Identifiers

CAS 7647-01-0, 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl
View More Specs

Products 6
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10133080
SDS View Documents
Thermo Scientific Acros
124635000
500 mL Glass Bottle
23.10 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10362913
SDS View Documents
Thermo Scientific Acros
124635001
500 mL Plastic bottle
23.62 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10794821
SDS View Documents
Thermo Scientific Acros
124630010
1 L Glass Bottle
33.57 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10478660
SDS View Documents
Thermo Scientific Acros
124630025
2.5 L Glass Bottle
57.69 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10776233
SDS View Documents
Thermo Scientific Acros
124630026
2.5 L Plastic bottle
58.89 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10755663
SDS View Documents
Thermo Scientific Acros
124630100
10 L Plastic drum
151.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrochloric acid, for analysis, fuming, 37% solution in water, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.