missing translation for 'onlineSavingsMsg'
Learn More
Hydrofluoric acid, for analysis, 48 to 51% solution in water, Thermo Scientific Chemicals Product Code.: 10286370

Hydrofluoric acid, for analysis, 48 to 51% solution in water, Thermo Scientific Chemicals

Product Code. 10286370
500 mL, Plastic bottle
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Plastic bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10286370

Brand: Thermo Scientific Acros 223335000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-39-3, 7732-18-5
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484Show MoreShow Less
PubChem CID 14917
ChEBI CHEBI:29228
IUPAC Name hydrogen fluoride
SMILES F

Specifications

Color Colorless
Melting Point -35.0°C
Boiling Point 105.0°C
Density 1.1500g/mL
Heavy Metals (as Pb) 0.5ppm max.
Packaging Plastic bottle
Residue after Ignition 0.0005% max.
Specific Gravity 1.15
Quantity 500 mL
Linear Formula HF
Physical Form Liquid
Fieser 06,284; 09,238; 16,176
Merck Index 15, 4827
Solubility Information Solubility in water: soluble.
Formula Weight 20
Percent Purity 48 to 51%
Grade Analytical
Chemical Name or Material Hydrofluoric acid, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463905 Certificate TypeCertificate of Analysis Date17/06/2024 Catalog Number223330250,223330025,223335000
Lot NumberA0460265 Certificate TypeCertificate of Analysis Date14/02/2024 Catalog Number223330250,223330025,223335000
Lot NumberA0441454 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number223330250,223330025,223335000
Lot NumberA0450069 Certificate TypeCertificate of Analysis Date03/02/2023 Catalog Number223330250,223330025,223335000
Lot NumberA0395221 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number223330250,223330025,223335000
compliance-icons compliance-icons
Product Identifier
  • Hydrofluoric acid, 48 to 51% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrofluoric acid, for analysis, 48 to 51% solution in water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.