Learn More
Thermo Scientific™ HyperSep™ Pressure Manifold Installation Kit
Obtain all the parts needed to set up your Thermo Scientific™ HyperSep™ Positive Pressure Manifold.
Brand:  Thermo Scientific™ 60104-272
Product Code. 13436926
  • 518.00 € / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use the Thermo Scientific™ HyperSep™ Positive Pressure Manifold Installation Kit for all the parts needed to set up your manifold. The HyperSep Positive Pressure Manifold holds up to 48 SPE cartridges. Samples and solvent, added to the SPE cartridge, are extracted under positive pressure for an even flow at each position. Extracts are collected into 13 or 16mm tubes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Installation Kit, 1 pk
Positive Pressure Manifold
Installation Kit
SPE
HyperSep
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ HyperSep™ Pressure Manifold Installation Kit > Installation Kit