Learn More
Invitrogen™ iBlot™ 3 Transfer Stacks, mini, nitrocellulose

iBlot 3 Transfer Stacks are used with iBlot 3 Western Blot Transfer Device for the transfer of proteins from an SDS-PAGE gel to a membrane support when performing western blot experiments.
Brand: Invitrogen™ IB33002
183.35 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
iBlot 3 Transfer Stacks are used with the iBlot 3 Western Blot Transfer Device for the transfer of proteins from an SDS-PAGE gel to a membrane support when performing western blot experiments. iBlot 3 Transfer Stacks contain copper-fiber electrode sheets, optimized cathode and anode solid buffer gels, and a preconditioned nitrocellulose transfer membrane. They are compatible with common commercially available protein gels, including Bis-Tris, Tris-Glycine, Tris-Acetate, and Tricine chemistries. This mini-size stack (8 x 8 cm) can be used with one mini-size gel.
The iBlot 3 Western Blot Transfer System (iBlot 3 Transfer Stacks plus iBlot 3 Transfer Device) has been optimized to provide protein transfers in 3–8 minutes that produce higher quality and more reliable western blot result compared to many other transfer methods.
Nitrocellulose membrane
The nitrocellulose membrane (0.2-μm pore size) in this stack is composed of 100% pure nitrocellulose to provide high-quality transfer. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. Proteins bind to the membrane due to hydrophobic and electrostatic interactions. The protein-binding capacity is 209 μg/cm2.

Specifications
8 cm | |
10 Stacks | |
Nitrocellulose | |
Store at room temperature. | |
iBlot | |
Less waste, sustainable packaging, sustainable disposal |
8 cm | |
Transfer Stack | |
0.2 μm | |
8 x 8 cm (Mini) | |
10 Blots |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3NM270423-01 | Certificate of Analysis | 12/02/2025 | |
3NM241124-01 | Certificate of Analysis | 12/02/2025 | |
3NM010424-01 | Certificate of Analysis | 12/02/2025 | |
3NM290523-01 | Certificate of Analysis | 12/02/2025 | |
3NM110724-01 | Certificate of Analysis | 12/02/2025 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.