missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Indole-6-carboxaldehyde, 97%

111.00 € - 333.00 €

Chemical Identifiers

CAS 1196-70-9
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD02179596
InChI Key VSPBWOAEHQDXRD-UHFFFAOYSA-N
Synonym indole-6-carboxaldehyde, 6-formylindole, 1h-indole-6-carboxaldehyde, indole-6-carbaldehyde, 6-indolecarboxaldehyde, indole-6-carboxalehyde, 6-formyl-1h-indole, indole-6-carboxaldehyde 6-formylindole, 6-formyl-indole, 6-formyl indole
PubChem CID 2773435
IUPAC Name 1H-indole-6-carbaldehyde
SMILES C1=CC(=CC2=C1C=CN2)C=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15431318
View Documents
Alfa Aesar™
H55152.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15421318
View Documents
Alfa Aesar™
H55152.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

1196-70-9
145.161
VSPBWOAEHQDXRD-UHFFFAOYSA-N
2773435
C1=CC(=CC2=C1C=CN2)C=O
C9H7NO
MFCD02179596
indole-6-carboxaldehyde, 6-formylindole, 1h-indole-6-carboxaldehyde, indole-6-carbaldehyde, 6-indolecarboxaldehyde, indole-6-carboxalehyde, 6-formyl-1h-indole, indole-6-carboxaldehyde 6-formylindole, 6-formyl-indole, 6-formyl indole
1H-indole-6-carbaldehyde

Specifications

1196-70-9
Brown to White
MFCD02179596
VSPBWOAEHQDXRD-UHFFFAOYSA-N
1H-indole-6-carbaldehyde
2773435
97%
Indole-6-carboxaldehyde
127°C to 131°C
C9H7NO
indole-6-carboxaldehyde, 6-formylindole, 1h-indole-6-carboxaldehyde, indole-6-carbaldehyde, 6-indolecarboxaldehyde, indole-6-carboxalehyde, 6-formyl-1h-indole, indole-6-carboxaldehyde 6-formylindole, 6-formyl-indole, 6-formyl indole
C1=CC(=CC2=C1C=CN2)C=O
145.161
145.16
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P363-P321-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash contaminated clothing before reuse.
Specific treatment (see ... on this label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.