Learn More
KARTELL Industrial Tank
Industrial Tanks are specifically designed for stacking to promote stability during transportation for industry and laboratory practice.
Brand: KARTELL 0178300
This item is not returnable.
View return policy
Description
- Standardized industrial packaging design, and even if specifically addressed to a use in industrial packaging, they could result useful in everyday laboratory practice.
- They are made of High Density PE food grade, with a comfortable handle, and feature a secure seal-cap following DIN recommendations.
- They are conform to UN regulation on transportation of dangerous and toxic liquids and are suitable for road (ADR-RID), sea (IMO-IMDG) and air (IATA-ICAO) transport.
- SUITABLE FOR FOODSTUFF.
Specifications
Included | |
Polyethylene | |
6 pcs. |
White | |
20 L |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
KARTELL Industrial Tank > Capacity: 20 L
Spot an opportunity for improvement?Share a Content Correction