Learn More
Jenway™ Air Tubing
Brand:  Jenway™ 10089-43
Product Code. 12778686
  • 28.55 € / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Model PFP7 or PFP7/C Flame photometer
Air tube (2 m)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Jenway™ Air Tubing > For Use With: For Atomiser PFP240