missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP

ATP, CTP, GTP, UTP (nucleoside 5'-triphosphates), and 10 mM NTP Mix are suitable for use in in vitro transcription reactions with SP6, T3, and T7 RNA polymerases for production of RNA probes, or RNA transcripts for translation in in vitro translation extracts.

Brand:  Invitrogen™ 18330019

Product Code. 10114912

  • 131.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ATP, CTP, GTP, UTP (nucleoside 5´-triphosphates), and 10 mM NTP Mix are suitable for use in in vitro transcription reactions with SP6, T3, and T7 RNA polymerases for production of RNA probes, or RNA transcripts for translation in in vitro translation extracts. These nucleotides are supplied as convenient, ready-to-use solutions at a concentration of 10 mM in 1 mM Tris-HCl (pH 7.5).

Related products:
• 10 mM CTP (Cat. No. 18331017)
• 10 mM GTP (18332015)
• 10 mM UTP (18333013)
• 10 mM NTP Mix (18109017)

Order Info

Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 mM ATP in Tris-HCl (500 μL)

Store at –5 to –30°C.
10 mM
500 μL
Liquid
Approved for shipment on Wet or Dry Ice
Unlabeled
in vitro Transcription
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3060776 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number18330019
Lot Number2477055 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number18330019
Lot Number2794106 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number18330019
Lot Number2840463 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number18330019
Lot Number2546995 Certificate TypeCertificate of Analysis Date18/03/2023 Catalog Number18330019
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ ATP > 5μmol

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.