missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ATP Solution (100 mM)

Thermo Scientific ATP (adenosine 5'-triphosphate) is an extremely stable nucleotide, and it is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH.

Brand:  Thermo Scientific™ R0441

40.72 EUR valid until 2025-12-31
Use promo code "24368" to get your promotional price.


Product Code. 10304340

  • 44.21 € / 0.25mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific ATP (adenosine 5'-triphosphate) is an extremely stable nucleotide, and it is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. This nucleotide is used in different molecular biology applications, and it has greater than 99% purity.

Features
• Greater than 99% purity confirmed by HPLC
• Functionally tested in in vitro transcription
• Stable for two years at -20°C and after multiple freeze-thaw cycles

Applications
in vitro transcription
• aRNA synthesis
• siRNA synthesis
• RNA amplification
• Ligation
• Phosphorylation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mM
ATP Solution, 100 mM (0.25 mL)

Store at –20°C.
Unconjugated
ATP Solution
in vitro Transcription, RNA Amplification
Dry Ice
0.25 mL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2812530 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberR0441
Lot Number2824059 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0441
Lot Number2817014 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0441
Lot Number2791890 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0441
Lot Number2822122 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0441
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ ATP Solution (100 mM) > 0.25mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.