missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™-Pro PCR Clean-Up Kit
GREENER_CHOICE

The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfere with enzymatic reactions.

Brand:  Invitrogen™ CS32250

 View more versions of this product

Product Code. 10430164

  • 796.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Kit contains columns, collection tubes, recovery tubes, binding buffer, wash buffer, and elution buffer.
This item is not returnable. View return policy

Description

Description

The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfere with enzymatic reactions.

• 100% ethanol-free, guanidinium-free, organic solvent-free protocols
• Improved downstream performance
• Complete kit for fast, hassle-free setup
• 20% fewer handling steps than most PCR clean-up protocols
• Purification of up to 500 μL of PCR product in one column

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

The ChargeSwitch™-Pro PCR Clean-up Kit contains columns, collection tubes, recovery tubes, binding buffer, wash buffer, and elution buffer.

Store at room temperature. Guaranteed stable for 6 months when properly stored.

Fewer resources used and less waste, Less hazardous
ChargeSwitch™
250 Preps
DNA from Enzymatic Reactions (e.g. PCR)
ChargeSwitch™ Chemistry
PCR Products
Kit
Not High-throughput Compatible (Manual)
PCR Clean-Up Kit
Room Temperature
Next-Generation Sequencing (NGS), Cloning, Standard PCR
250 Preps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2209798 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberCS32250
Lot Number2575812 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberCS32250
Lot Number2595535 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberCS32250
Lot Number2631800 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberCS32250
Lot Number2926478 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberCS32250
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ ChargeSwitch™-Pro PCR Clean-Up Kit > 250 preps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.