missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Luria Broth Base (Miller's LB Broth Base), powder

Luria broth (LB) is a popular, rich medium used to cultivate bacteria and to perform coliphage plaque assays.

Brand:  Invitrogen™ 12795027

82.95 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 11548866

  • 105.00 € / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Luria broth (LB) is a popular, rich medium used to cultivate bacteria and to perform coliphage plaque assays. The original formulation published in 1951 by Guiseppe Bertani to study phage plaque formation and named Lysogeny broth. It is also known as Lennox broth and Luria-Bertani broth.

Widely used LB medium formulations differ in salt concentration:
Miller: 10 g/L NaCl
Lennox: 5 g/L NaCl
Luria: 0.5 g/L NaCl

Luria Broth Base (Miller's formulation) is recommended for the general maintenance and growth of E. coli as it allows for fast cell expansion and supports high cell density. In cases where salt-sensitive antibiotics are being used for selection (such as Zeocin, tetracycline or chloramphenicol), Lennox formulation medium would be recommended.

Luria Broth Base formulation
Formulation per liter: 10 g SELECT Peptone 140, 5 g SELECT Yeast Extract, and 10 g NaCl.
To prepare 1 L of Miller-LB media, add 25 g of powder to 1 L distilled water and mix by vortexing. Adjust the pH to 7.0 with 5 N NaOH and sterilize by autoclaving for 25 minutes at 121°C.

Find the media format that fits your needs
We offer other ready-to-use media formulations, such as LB Broth (Cat. No. 10855001), Terrific Broth (Cat. No. A1374301) or S.O.C recovery medium (Cat. No. 15544034).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Escherichia coli
Bottle
Autoclave
Coliforms
Room Temperature
• 500 g LB Dehydrated Media Powder

Store at room temperature.

LB Broth Base (Miller's LB Broth Base)
Dehydrated Culture Media
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2407AA Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number12795027
Lot Number2408DL Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number12795027
Lot Number591021G007 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12795027
Lot Number591021G009 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12795027
Lot Number2208XX Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12795027
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Luria Broth Base (Miller's LB Broth Base), powder > 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.