missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCR SuperMix

PCR SuperMix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification.

Brand:  Invitrogen™ 10572014

Product Code. 10093152

  • 257.00 € / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

PCR SuperMix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification. Simply add template and primers, reducing set-up time by half. PCR SuperMix contains Mg2+, dNTPs, and recombinant Taq DNA Polymerase at concentrations sufficient for routine PCR of fragments up to 5 kb. PCR SuperMix is supplied at a 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions.

For superior PCR performance, we recommend DreamTaq DNA Polymerase.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 4 vials of PCR SuperMix, 1.125 mL each

Store at -20°C.
Low
Standard
PCR SuperMix
Wet or Dry Ice
1.1X
No
3'-A
5 kb or less
Tube
Taq DNA Polymerase
5' - 3'
100 rxns
Standard PCR
1X
100 Reactions
SuperMix or Master Mix
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3040491 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number10572014
Lot Number2448821 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10572014
Lot Number2477602 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10572014
Lot Number2529653 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10572014
Lot Number2596682 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10572014
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ PCR SuperMix > 100 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.