Learn More
Invitrogen™ SuperScript™ III Platinum™ One-Step qRT-PCR Kit
Combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient one-step format.
Brand: Invitrogen™ 11732020
373.00 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
381.00 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Includes: 100μL SuperScript III/Platinum Taq Mix; 2 x 1.25mL 2X Reaction Mix; 1mL Magnesium Sulfate (50mM); 100μL ROX Reference Dye (25μM)
Description
SuperScript™ III Platinum™ One-Step qRT-PCR Kit provides sensitive, specific detection with fluorogenic primers and probes, while the SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit offers the ease and convenience of SYBR™ Green I detection.
- The SuperScript III Platinum One-Step qRT-PCR Kit’s proven formula delivers reliable, accurate target detection when it matters most
- A convenient, one-step format and consistent performance make it ideal for a number of gene expression applications, including infectious disease surveillance
- The kit has been validated by several global public health organizations and has long been the choice of the US CDC for infectious disease surveillance
- SuperScript III RT for higher temperature cDNA synthesis when addressing difficult RNA secondary structure
- Platinum Taq DNA Polymerase with hot-start technology for improved specificity
- LUX fluorogenic primers and dual-labeled fluorogenic probes for effective detection performance
- Validated performance on multiple real-time instrument platforms, including rotor-based systems
- Multiple SuperScript III Platinum One-Step qRT-PCR Kit configurations are available
- Store components at -20°C, ROX Reference Dye must be stored in the dark
Order Info
Shipping Condition: Dry ice

Specifications
• 100 μl SuperScript™ III/Platinum™ Taq Mix • 2 × 1.25 ml 2X Reaction Mix • 1 ml Magnesium Sulfate (50 mM) • 100 μl ROX Reference Dye (25 μM) Store components at -20°C. ROX Reference Dye must be stored in the dark. |
|
1-step RT-qPCR | |
Standard | |
Platinum™, SuperScript™ | |
100 reactions | |
100 Reactions | |
100 Reactions |
Primer-probe | |
Taq DNA Polymerase | |
7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ | |
One-Step qRT-PCR Kit | |
Dry Ice | |
Gene Expression, Pathogen Detection, Virus Detection | |
RNA |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2998002 | Certificate of Analysis | 31/01/2025 | 11732020 |
3120259 | Certificate of Analysis | 31/01/2025 | 11732020 |
3057273 | Certificate of Analysis | 30/01/2025 | 11732020 |
2427130 | Certificate of Analysis | 29/01/2025 | 11732020 |
2443930 | Certificate of Analysis | 29/01/2025 | 11732020 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.