missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Proteinase K Solution (20 mg/mL), RNA grade

Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.

Brand:  Invitrogen™ 25530049

344.85 EUR valid until 2025-12-31
Use promo code "24274" to get your promotional price.



Product Code. 11588916

  • 394.00 € / 100mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.

  • Over a wide pH range—optimal activity between 6.5 and 9.5
  • Under denaturing conditions—e.g., in the presence of SDS or urea
  • In the presence of metal chelating agents—e.g., EDTA
  • At comparatively high temperatures—optimum digestion temperature is 65°C
  • Protein Filter: P

Applications
Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.

Performance and quality testing
Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Unit definition
One mAnson unit is described as that amount of enzyme that liberates 1 μmol of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

20 mg/mL
Chromatin Biology/RNA Extraction/DNA Extraction
Approved for shipment on Wet or Dry Ice
Proteinase K
DNase-Free, RNase-Free
Store in freezer (-5 to -30°C).
5 mL
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2443899 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number25530049
Lot Number2449367 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number25530049
Lot Number2481060 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number25530049
Lot Number2549630 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number25530049
Lot Number2449370 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number25530049
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Proteinase K Solution (20 mg/mL), RNA grade > 5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.