Learn More
Invitrogen™ SYBR™ Green I Nucleic Acid Gel Stain, 10,000X concentrate in DMSO
Extremely sensitive stain for detecting double-stranded DNA (dsDNA) in agarose and polyacrylamide gels
Brand: Invitrogen™ S7585
Description
SYBR Green I offers greater sensitivity for dsDNA, making it especially useful for assays where the presence of contaminating RNA or ssDNA might obscure results. With exceptionally low background fluorescence and spectral characteristics that closely match light sources and filter sets in existing instruments, SYBR Green I stain is ideal for use with laser scanners.
Product use 1mL stains 100 minigels.
- Sensitivity: at least four times greater than ethidium bromide for DNA in agarose gels
- Ease of use: gels soaked in diluted stain can be visualized without desalting
- Compatibility: with UV transilluminators, gel documentation systems, and laser scanners
- Flexibility: for use with a broad range of applications, including DNA typing, PCR-based assays, DNA damage assays, analysis of complex samples, and real-time PCR detection
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room temperature

Specifications
• Supplied as a 10,000X concentrate in DMSO Store at -20°C, protected from light in a dessicator. |
|
Fluorescence | |
Nucleic Acid Gel Stain | |
Room Temperature |
In-Gel Detection | |
SYBR Green I | |
20 x 50 μL | |
DNA |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.