missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR™ Green I Nucleic Acid Gel Stain, 10,000X concentrate in DMSO

Extremely sensitive stain for detecting double-stranded DNA (dsDNA) in agarose and polyacrylamide gels

Brand:  Invitrogen™ S7563

 View more versions of this product

Product Code. 10710004

  • 578.00 € / 500µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

SYBR Green I offers greater sensitivity for dsDNA, making it especially useful for assays where the presence of contaminating RNA or ssDNA might obscure results. With exceptionally low background fluorescence and spectral characteristics that closely match light sources and filter sets in existing instruments, SYBR Green I stain is ideal for use with laser scanners.
Product use 1mL stains 100 minigels.

  • Sensitivity: at least four times greater than ethidium bromide for DNA in agarose gels
  • Ease of use: gels soaked in diluted stain can be visualized without desalting
  • Compatibility: with UV transilluminators, gel documentation systems, and laser scanners
  • Flexibility: for use with a broad range of applications, including DNA typing, PCR-based assays, DNA damage assays, analysis of complex samples, and real-time PCR detection

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Supplied as a 10,000X concentrate in DMSO

Store at -20°C, protected from light in a dessicator.
Fluorescence
Nucleic Acid Gel Stain
Room Temperature
In-Gel Detection
SYBR Green I
500 μL
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2720336 Certificate TypeCertificate of Analysis Date06/11/2023 Catalog NumberS7563
Lot Number2533897 Certificate TypeCertificate of Analysis Date07/10/2022 Catalog NumberS7563
Lot Number1135054 Certificate TypeCertificate of Analysis Date23/12/2021 Catalog NumberS7563
Lot Number2415757 Certificate TypeCertificate of Analysis Date21/12/2021 Catalog NumberS7563
Lot Number2311857 Certificate TypeCertificate of Analysis Date20/05/2021 Catalog NumberS7563
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ SYBR™ Green I Nucleic Acid Gel Stain, 10,000X concentrate in DMSO > 500μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.