Learn More
Invitrogen™ T4 DNA Ligase (1 U/μL)
Ideal for cloning (blunt-end or cohesive-end ligation) and adding linkers or adapters to blunt-ended DNA
Brand: Invitrogen™ 15224025
251.22 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Includes: Vial of 5X reaction buffer (250mM Tris-HCl (pH 7.6), 50mM MgCl2, 5mM ATP, 5mM DTT, 25% (w/v) polyethylene glycol-8000)
Description
T4 DNA Ligase catalyzes the formation of phosphodiester bonds in the presence of ATP between double-stranded DNAs with 3'hydroxyl and 5'phosphate termini. Single-stranded nucleic acids are not substrates for this enzyme.
- Optimizes ligation in 5 minutes
- Source: Purified from E. coli lambda lysogen NM989
- Performance and quality testing: Endodeoxyribonuclease, 3' and 5' exodeoxyribonuclease assays; ligation efficiency tested
- Unit definition: One unit catalyzes exchange of 1nmol 32P-labeled pyrophosphate into ATP in 20 min. at 37°C. One unit is equal to approximately 300 cohesive-end ligation units
- Unit reaction conditions: 66mM Tris-HCl (pH 7.6), 6.6mM MgCl2, 10mM DTT, 66μM ATP, 3.3μM 32P-labeled pyrophosphate, and enzyme in 0.1mL for 20 min. at 37°C
ChIP-on-Chip, Chromatin Biology, Cloning, RNAi, Epigenetics & Noncoding RNA Research, Restriction Enzyme Cloning
Order Info
Shipping Conditions: Dry ice

Specifications
1 U/μL | |
Dry Ice | |
5X Reaction Buffer | |
T4 DNA Ligase |
T4 DNA Ligase is supplied with a vial of 5X reaction buffer [250 mM Tris-HCl (pH 7.6), 50 mM MgCl2 , 5 mM ATP, 5 mM DTT, 25% (w/v) polyethylene glycol-8000]. Store at -20°C. | |
Ligase | |
500 U |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3031569 | Certificate of Analysis | 31/01/2025 | 15224025 |
3021986 | Certificate of Analysis | 31/01/2025 | 15224025 |
2482358 | Certificate of Analysis | 29/01/2025 | 15224025 |
2509781 | Certificate of Analysis | 29/01/2025 | 15224025 |
2556510 | Certificate of Analysis | 29/01/2025 | 15224025 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.