Learn More
Invitrogen™ T4 DNA Ligase (1 U/μL)
Ideal for cloning (blunt-end or cohesive-end ligation) and adding linkers or adapters to blunt-ended DNA
70.50 € - 986.00 €
Specifications
Concentration | 1 U/μL |
---|---|
Enzyme | Ligase |
Product Type | T4 DNA Ligase |
Includes: Vial of 5X reaction buffer (250mM Tris-HCl (pH 7.6), 50mM MgCl2, 5mM ATP, 5mM DTT, 25% (w/v) polyethylene glycol-8000)
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10124752
|
Invitrogen™
15224017 |
100 U |
70.50 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10534613
|
Invitrogen™
15224025 |
500 U |
318.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11558726
|
Invitrogen™
15224090 |
4 x 500 U |
986.00 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
T4 DNA Ligase catalyzes the formation of phosphodiester bonds in the presence of ATP between double-stranded DNAs with 3'hydroxyl and 5'phosphate termini. Single-stranded nucleic acids are not substrates for this enzyme.
- Optimizes ligation in 5 minutes
- Source: Purified from E. coli lambda lysogen NM989
- Performance and quality testing: Endodeoxyribonuclease, 3' and 5' exodeoxyribonuclease assays; ligation efficiency tested
- Unit definition: One unit catalyzes exchange of 1nmol 32P-labeled pyrophosphate into ATP in 20 min. at 37°C. One unit is equal to approximately 300 cohesive-end ligation units
- Unit reaction conditions: 66mM Tris-HCl (pH 7.6), 6.6mM MgCl2, 10mM DTT, 66μM ATP, 3.3μM 32P-labeled pyrophosphate, and enzyme in 0.1mL for 20 min. at 37°C
ChIP-on-Chip, Chromatin Biology, Cloning, RNAi, Epigenetics & Noncoding RNA Research, Restriction Enzyme Cloning
Order Info
Shipping Conditions: Dry ice
Specifications
1 U/μL | |
Ligase | |
T4 DNA Ligase |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.