missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ T4 DNA Ligase (1 U/μL)

Ideal for cloning (blunt-end or cohesive-end ligation) and adding linkers or adapters to blunt-ended DNA

Brand:  Invitrogen™ 15224090

778.94 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 11558726

  • 986.00 € / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Vial of 5X reaction buffer (250mM Tris-HCl (pH 7.6), 50mM MgCl2, 5mM ATP, 5mM DTT, 25% (w/v) polyethylene glycol-8000)
This item is not returnable. View return policy

Description

Description

T4 DNA Ligase catalyzes the formation of phosphodiester bonds in the presence of ATP between double-stranded DNAs with 3'hydroxyl and 5'phosphate termini. Single-stranded nucleic acids are not substrates for this enzyme.

  • Optimizes ligation in 5 minutes
  • Source: Purified from E. coli lambda lysogen NM989
  • Performance and quality testing: Endodeoxyribonuclease, 3' and 5' exodeoxyribonuclease assays; ligation efficiency tested
  • Unit definition: One unit catalyzes exchange of 1nmol 32P-labeled pyrophosphate into ATP in 20 min. at 37°C. One unit is equal to approximately 300 cohesive-end ligation units
  • Unit reaction conditions: 66mM Tris-HCl (pH 7.6), 6.6mM MgCl2, 10mM DTT, 66μM ATP, 3.3μM 32P-labeled pyrophosphate, and enzyme in 0.1mL for 20 min. at 37°C

ChIP-on-Chip, Chromatin Biology, Cloning, RNAi, Epigenetics & Noncoding RNA Research, Restriction Enzyme Cloning

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 U/μL
Ligase
4 x 500 U
T4 DNA Ligase is supplied with a vial of 5X reaction buffer [250 mM Tris-HCl (pH 7.6), 50 mM MgCl2 , 5 mM ATP, 5 mM DTT, 25% (w/v) polyethylene glycol-8000]. Store at -20°C.
5X Reaction Buffer
T4 DNA Ligase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ T4 DNA Ligase (1 U/μL) > 4 x 500 units

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.