Learn More
Invitrogen™ Total Exosome Isolation Reagent (from cell culture media)
Total Exosome Isolation Reagent (from cell culture media) enables fast and efficient enrichment of intact exosomes from cell culture media samples.
Brand: Invitrogen™ 4478359
358.70 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Total Exosome Isolation Reagent (from cell culture media) enables fast and efficient enrichment of intact exosomes from cell culture media samples.
- Maximize recovery of intact exosomes for any type of downstream application
- Maximize flexibility—can be scaled up or down depending on the sample size
- Enables simple and reliable concentration of intact exosomes from cell media samples
Order Info
Shipping Condition: Room Temperature

Specifications
Store at 2°C to 8°C. | |
Exosomes | |
Cell Culture Media |
Total Exosome Isolation | |
50 mL | |
Room Temperature |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2752563 | Certificate of Analysis | 28/01/2025 | 4478359 |
2811377 | Certificate of Analysis | 28/01/2025 | 4478359 |
2879178 | Certificate of Analysis | 26/01/2025 | 4478359 |
2855494 | Certificate of Analysis | 26/01/2025 | 4478359 |
2718592 | Certificate of Analysis | 26/01/2025 | 4478359 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.