missing translation for 'onlineSavingsMsg'
Learn More

Iodine concentrate, For 1L standard solution, 0.05 M I2 (0.1 N), Honeywell Fluka™

Product Code. 15675200
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.

for 1L standard solution, 0.05 M I2 (0.1 N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0, 7553-56-2, 7732-18-5

Specifications

Quantity 1 Pc.
Molecular Formula I2
MDL Number MFCD00011355 MFCD00164163
UN Number NONH for all modes of transport
Beilstein 3587194
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES II
IUPAC Name diiodine
Molecular Weight (g/mol) 253.81
PubChem CID 807
ChEBI CHEBI:17606
Formula Weight 253.81g/mol
Concentration or Composition (by Analyte or Components) 0.05M
Packaging Ampoule
Chemical Name or Material Iodine concentrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodine concentrate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : iodine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'