missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodoacetic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
50 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 79.0°C to 83.0°C |
Color | Brown to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Linear Formula | ICH2CO2H |
Quantity | 25 g |
Beilstein | 02, 222 |
Merck Index | 15, 5068 |
Solubility Information | Solubility in water: 600g/L (20 c). Other solubilities: soluble in alcohol,slightly soluble in ether |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0463407 | Certificate of Analysis | 06/06/2024 | 122280500,122281000,122280250 |
A0436287 | Certificate of Analysis | 25/08/2023 | 122280500,122281000,122280250 |
A0449166 | Certificate of Analysis | 13/01/2023 | 122280500,122281000,122280250 |
A0401484 | Certificate of Analysis | 15/12/2022 | 122280500,122281000,122280250 |
A0415190 | Certificate of Analysis | 13/12/2022 | 122280500,122281000,122280250 |
Safety and Handling


Product Identifier
- Iodoacetic acid
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Iodoacetic acid
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Iodoacetic acid, 97%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction